Learn More
Biolegend PE anti-mouse CD45
Supplier:  Biolegend 147712
Encompass_Preferred
PE anti-mouse CD45 [I3/2.3]; Isotype: Rat IgG2b; Reactivity: Mouse; Apps: FC; Size: 200 ug
Catalog No. 50-711-942