Learn More
Biolegend PE anti-mouse CD85k (gp49 Receptor)
Supplier:  Biolegend 144904
Encompass_Preferred
PE anti-mouse CD85k (gp49 Receptor) [H1.1]; Isotype: Armenian Hamster IgG; Reactivity: Mouse; Apps: FC; Size: 100 ug
Catalog No. NC1282936