Learn More
Biolegend PE anti-mouse CD90 Thy1
Supplier:  Biolegend 166407
Encompass_Preferred
PE anti-mouse CD90 (Thy1) [W20280E]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 25 μg
Catalog No. 50-286-5968