Learn More
Biolegend PE anti-mouse CD95 (Fas)
Supplier:  Biolegend 152608
Encompass_Preferred
PE anti-mouse CD95 (Fas) [SA367H8]; Isotype: Mouse IgG1, κ; Reactivity: Mouse; Apps: FC; Size: 100 ug
Catalog No. NC1791094