Learn More
Biolegend PE anti-mouse Folate Receptor β (FR-β)
Supplier:  Biolegend 153304
Encompass_Preferred
PE anti-mouse Folate Receptor β (FR-β) [10/FR2]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 100 ug
Catalog No. 50-207-2117