Learn More
Biolegend PE anti-mouse IL-23R
Supplier:  Biolegend 150903
Encompass_Preferred
PE anti-mouse IL-23R [12B2B64]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC; Size: 25 ug
Catalog No. NC1465725