Learn More
Biolegend PE anti-mouse IRF5
Supplier:  Biolegend 158603
Encompass_Preferred
PE anti-mouse IRF5 [W16007B]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: ICFC; Size: 25 ug
Catalog No. 50-207-2400