Learn More
Biolegend PE anti-mouse P2X7R
Supplier:  Biolegend 148703
Encompass_Preferred
PE anti-mouse P2X7R [1F11]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC; Size: 25 ug
Catalog No. 50-207-1812