Learn More
Cytek Biosciences Inc Antibodies PE Anti-Mouse TER-119 (TER-119) ..... Cat. No. 50-5921-U025 ..... Size 25 µg
Supplier:  Cytek Biosciences Inc 505921U025
Encompass_Preferred
Antibodies PE Anti-Mouse TER-119 (TER-119) ..... Cat. No. 50-5921-U025 ..... Size 25 µgThe TER-119 antibody is named for the antigen to which it binds, a 52 kDa surface protein that is associated with glycophorin-A. TER-119 is considered to be a lineage marker for later stages of erythroid cell development, as its expression begins at the pro-erythroblast stage. TER-119 antigen is not expressed at either BFU-E or CFU-E stages, i.e. prior to the pro-erythroblast stage.
Catalog No. 50-210-3443