Learn More
Cytek Biosciences Inc Antibodies PE Anti-Mouse TIM-4 (RMT4-54) ..... Cat. No. 50-5866-U025 ..... Size 25 µg
Supplier:  Cytek Biosciences Inc 505866U025
Encompass_Preferred
Antibodies PE Anti-Mouse TIM-4 (RMT4-54) ..... Cat. No. 50-5866-U025 ..... Size 25 µgThe RMT4-54 antibody reacts with mouse Tim-4, also known as T cell immunoglobulin and mucin domain containing protein-4. Tim-4 is expressed on macrophages and dendritic cells and can serve as a receptor for Tim-1. Tim-4 also functions as a receptor for phosphatidylserine and serves to enhance the phagocytosis of apoptotic cells.
Catalog No. 50-210-3436