Learn More
Biolegend PE anti-mouse CD371 (CLEC12A)
Supplier:  Biolegend 143403
Encompass_Preferred
PE anti-mouse CD371 (CLEC12A) [5D3/CLEC12A]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 25 ug
Catalog No. NC1905973