Learn More
Biolegend PE anti-mouse CD105
Supplier:  Biolegend 120408
Encompass_Preferred
PE anti-mouse CD105 [MJ7/18]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 100 ug
Catalog No. 50-163-857