Learn More
Biolegend PE anti-mouse CD107b (Mac-3)
Supplier:  Biolegend 108505
Encompass_Preferred
PE anti-mouse CD107b (Mac-3) [M3/84]; Isotype: Rat IgG1, κ; Reactivity: Mouse; Apps: FC; Size: 50 ug
Catalog No. 50-163-271