Learn More
Biolegend PE anti-mouse CD123
Supplier:  Biolegend 106005
Encompass_Preferred
PE anti-mouse CD123 [5B11]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 25 ug
Catalog No. 50-163-112