Learn More
Biolegend PE anti-mouse CD124 (IL-4Rα)
Supplier:  Biolegend 144804
Encompass_Preferred
PE anti-mouse CD124 (IL-4Rα) [I015F8]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC; Size: 100 ug
Catalog No. 50-605-106