Learn More
Biolegend PE anti-mouse CD127 (IL-7Rα)
Supplier:  Biolegend 135009
Encompass_Preferred
PE anti-mouse CD127 (IL-7Rα) [A7R34]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 50 ug
Catalog No. 50-164-789