Learn More
Biolegend PE anti-mouse CD132 (common γ chain)
Supplier:  Biolegend 132305
Encompass_Preferred
PE anti-mouse CD132 (common γ chain) [TUGm2]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC; Size: 50 ug
Catalog No. 50-164-655