Learn More
Biolegend PE anti-mouse CD133
Supplier:  Biolegend 141203
Encompass_Preferred
PE anti-mouse CD133 [315-2C11]; Isotype: Rat IgG2a, λ; Reactivity: Mouse; Apps: FC; Size: 25 ug
Catalog No. 50-165-186