Learn More
Biolegend PE anti-mouse CD152
Supplier:  Biolegend 106306
Encompass_Preferred
PE anti-mouse CD152 [UC10-4B9]; Isotype: Armenian Hamster IgG; Reactivity: Mouse; Apps: FC; Size: 200 ug
Catalog No. NC9038062