Learn More
Biolegend PE anti-mouse CD153
Supplier:  Biolegend 106405
Encompass_Preferred
PE anti-mouse CD153 [RM153]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC; Size: 50 ug
Catalog No. 50-163-129