Learn More
Biolegend PE anti-mouse CD178 (FasL)
Supplier:  Biolegend 106605
Encompass_Preferred
PE anti-mouse CD178 (FasL) [MFL3]; Isotype: Armenian Hamster IgG; Reactivity: Mouse; Apps: FC; Size: 50 ug
Catalog No. 50-163-136