Learn More
Biolegend PE anti-mouse CD194 (CCR4)
Supplier:  Biolegend 131203
Encompass_Preferred
PE anti-mouse CD194 (CCR4) [2G12]; Isotype: Armenian Hamster IgG; Reactivity: Mouse; Apps: FC; Size: 25 ug
Catalog No. 50-164-610