Learn More
Biolegend PE anti-mouse CD25
Supplier:  Biolegend 101904
Encompass_Preferred
PE anti-mouse CD25 [3C7]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC; Size: 100 ug
Catalog No. 50-162-606