Learn More
Biolegend PE anti-mouse CD254 (TRANCE, RANKL)
Supplier:  Biolegend 510005
Encompass_Preferred
PE anti-mouse CD254 (TRANCE, RANKL) [IK22/5]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 50 ug
Catalog No. 50-170-074