Learn More
Biolegend PE anti-mouse CD26 (DPP-4)
Supplier:  Biolegend 137804
Encompass_Preferred
PE anti-mouse CD26 (DPP-4) [H194-112]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 100 ug
Catalog No. 50-164-997