Learn More
Biolegend PE anti-mouse CD272 (BTLA)
Supplier:  Biolegend 139107
Encompass_Preferred
PE anti-mouse CD272 (BTLA) [6A6]; Isotype: Armenian Hamster IgG; Reactivity: Mouse; Apps: FC; Size: 25 ug
Catalog No. 50-207-1264