Learn More
Biolegend PE anti-mouse CD274 (B7-H1, PD-L1)
Supplier:  Biolegend 124307
Encompass_Preferred
PE anti-mouse CD274 (B7-H1, PD-L1) [10F.9G2]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC; Size: 25 ug
Catalog No. 50-164-174