Learn More
Biolegend PE anti-mouse CD279 (PD-1)
Supplier:  Biolegend 109103
Encompass_Preferred
PE anti-mouse CD279 (PD-1) [RMP1-30]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC; Size: 50 ug
Catalog No. 50-163-333