Learn More
Biolegend PE anti-mouse CD30
Supplier:  Biolegend 102306
Encompass_Preferred
PE anti-mouse CD30 [mCD30.1]; Isotype: Armenian Hamster IgG; Reactivity: Mouse; Apps: FC; Size: 200 ug
Catalog No. 50-162-674