Learn More
Biolegend PE anti-mouse CD309 (VEGFR2, Flk-1)
Supplier:  Biolegend 121905
Encompass_Preferred
PE anti-mouse CD309 (VEGFR2, Flk-1) [89B3A5]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 25 ug
Catalog No. 50-163-985