Learn More
Biolegend PE anti-mouse CD326 (Ep-CAM)
Supplier:  Biolegend 118206
Encompass_Preferred
PE anti-mouse CD326 (Ep-CAM) [G8.8]; Isotype: Rat IgG2a, κ; Reactivity: mouse; Apps: FC; Size: 200 ug
Catalog No. NC9994289