Learn More
Biolegend PE anti-mouse CD34
Supplier:  Biolegend 119308
Encompass_Preferred
PE anti-mouse CD34 [MEC14.7]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 200 ug
Catalog No. 50-163-793