Learn More
Biolegend PE anti-mouse CD355 (CRTAM)
Supplier:  Biolegend 142005
Encompass_Preferred
PE anti-mouse CD355 (CRTAM) [11-5/CRTAM]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 25 ug
Catalog No. 50-165-240