Learn More
Biolegend PE anti-mouse CD36
Supplier:  Biolegend 102605
Encompass_Preferred
PE anti-mouse CD36 [HM36]; Isotype: Armenian Hamster IgG; Reactivity: Mouse; Apps: FC; Size: 50 ug
Catalog No. 50-162-710