Learn More
Biolegend PE anti-mouse CD366 (Tim-3)
Supplier:  Biolegend 119704
Encompass_Preferred
PE anti-mouse CD366 (Tim-3) [RMT3-23]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 200 ug
Catalog No. 50-207-0367