Learn More
Biolegend PE anti-mouse CD4
Supplier:  Biolegend 116006
Encompass_Preferred
PE anti-mouse CD4 [RM4-4]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC; Size: 200 ug
Catalog No. 50-163-600