Learn More
Biolegend PE anti-mouse CD49d
Supplier:  Biolegend 103607
Encompass_Preferred
PE anti-mouse CD49d [R1-2]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC; Size: 50 ug
Catalog No. 50-162-865