Learn More
Biolegend PE anti-mouse CD62L
Supplier:  Biolegend 104408
Encompass_Preferred
PE anti-mouse CD62L [MEL-14]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 200 ug
Catalog No. 50-162-915