Learn More
Biolegend PE anti-mouse CD64 (FcγRI)
Supplier:  Biolegend 139303
Encompass_Preferred
PE anti-mouse CD64 (FcγRI) [X54-5/7.1]; Isotype: Mouse IgG1, κ; Reactivity: Mouse; Apps: FC; Size: 25 ug
Catalog No. 50-165-080