Learn More
Biolegend PE anti-mouse CD70
Supplier:  Biolegend 104606
Encompass_Preferred
PE anti-mouse CD70 [FR70]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC; Size: 200 ug
Catalog No. 50-162-961