Learn More
Biolegend PE anti-mouse CD80
Supplier:  Biolegend 104708
Encompass_Preferred
PE anti-mouse CD80 [16-10A1]; Isotype: Armenian Hamster IgG; Reactivity: Mouse, Cross-Reactivity: Dog (Canine); Apps: FC; Size: 200 ug
Catalog No. NC9895938