Learn More
Biolegend PE anti-mouse CD90.2 (Thy1.2)
Supplier:  Biolegend 140308
Encompass_Preferred
PE anti-mouse CD90.2 (Thy1.2) [53-2.1]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 200 ug
Catalog No. 50-165-126