Learn More
Biolegend PE anti-mouse FR4 (Folate Receptor 4)
Supplier:  Biolegend 125007
Encompass_Preferred
PE anti-mouse FR4 (Folate Receptor 4) [12A5]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC; Size: 25 ug
Catalog No. 50-164-229