Learn More
Biolegend PE anti-mouse GM-CSF
Supplier:  Biolegend 505405
Encompass_Preferred
PE anti-mouse GM-CSF [MP1-22E9]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: ICFC; Size: 25 ug
Catalog No. 50-169-900