Learn More
Biolegend PE anti-mouse/human CD207 (Langerin)
Supplier:  Biolegend 144204
Encompass_Preferred
PE anti-mouse/human CD207 (Langerin) [4C7]; Isotype: Mouse IgG2a, κ; Reactivity: Mouse, Human, Cross-Reactivity: Cynomolgus, Rabbit, Swine (Pig, Porcine); Apps: FC, IHC; Size: 100 ug
Catalog No. 50-403-152