Learn More
Biolegend PE anti-mouse I-Ab
Supplier:  Biolegend 116408
Encompass_Preferred
PE anti-mouse I-Ab [AF6-120.1]; Isotype: Mouse (BALB/c) IgG2a, κ; Reactivity: Mouse b haplotype, cross-reacts with H-2k and H-2u haplotypes; Apps: FC; Size: 200 ug
Catalog No. 50-163-651