Learn More
Biolegend PE anti-mouse IgM
Supplier:  Biolegend 406508
Encompass_Preferred
PE anti-mouse IgM [RMM-1]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 200 ug
Catalog No. 50-169-155