Learn More
Biolegend PE anti-mouse IL-12/IL-23 p40
Supplier:  Biolegend 505204
Encompass_Preferred
PE anti-mouse IL-12/IL-23 p40 [C15.6]; Isotype: Rat IgG1, κ; Reactivity: Mouse, IL-12/IL-23 p40 subunit (monomer, homodimer and heterodimer IL-12 p35/p40 or IL-23 p19/p40); Apps: ICFC; Size: 100 ug
Catalog No. 50-169-886