Learn More
Biolegend PE anti-mouse IL-1α
Supplier:  Biolegend 503203
Encompass_Preferred
PE anti-mouse IL-1α [ALF-161]; Isotype: Armenian Hamster IgG; Reactivity: Mouse; Apps: ICFC; Size: 25 ug
Catalog No. 50-169-790