Learn More
Biolegend PE anti-mouse Ly-51
Supplier:  Biolegend 108308
Encompass_Preferred
PE anti-mouse Ly-51 [6C3]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 200 ug
Catalog No. 50-163-240